ChargeSwitch™-Pro PCR Clean-Up Kit
Inquire about OEM or Commercial Supply version of this product here.
Product Image
Invitrogen™

ChargeSwitch™-Pro PCR Clean-Up Kit

Green features
The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfereRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
CS32250250 Preps
CS3205050 Preps
Catalog number CS32250
Price (USD)/ 250 preps
718.00
-
Add to cart
No. of Reactions:
250 Preps
Price (USD)/ 250 preps
718.00
Add to cart
ChargeSwitch™-Pro PCR Clean-Up Kit
Catalog numberCS32250
Price (USD)/ 250 preps
718.00
-
Add to cart

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

The ChargeSwitch™-Pro Kit provides:
• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column


How It Works
ChargeSwitch™ technology employs a completely novel mechanism that replaces harsh reagents such as chaotropic salts, ethanol, or organic solvents and isopropanol precipitation with a simple ion-exchange procedure. In this method, DNA binds to a surface ligand that is positively charged at low pH, and is eluted when the pH is raised to 8.5 (Figure 1). The ChargeSwitch™-Pro protocol retains the ease and familiarity of common silica spin-column protocols with binding, washing, and elution steps (Figure 2).


Better Downstream Results
Since ChargeSwitch™ technology does not require the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification, the resulting DNA is free of enzymatic inhibitors and thus shows better downstream performance (Figure 3). These compounds are known to inhibit many enzymatic reactions such as PCR, cloning, restriction enzyme digestion, and array CGH labeling. Expect to see reduced variability in your sequencing applications, with reads in excess of 800 bp with Phred scores >20 (Figure 4); clean restriction enzyme digestions (Figure 5); and removal of dNTPs and primers in excess of 98% (Figure 6).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
Green FeaturesFewer resources used and less waste, Less hazardous
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineChargeSwitch™
Product TypePCR Clean-Up Kit
Quantity250 Preps
Shipping ConditionRoom Temperature
TargetDNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application)Next-Generation Sequencing (NGS), Cloning, Standard PCR
Isolation TechnologyChargeSwitch™ Chemistry
No. of Reactions250 Preps
Sample TypePCR Products
Unit Size250 preps
Contents & Storage

The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

Store at room temperature. Guaranteed stable for 6 months when properly stored.