μPAC™ Neo Low-Load Trapping Columns
μPAC™ Neo Low-Load Trapping Columns
Thermo Scientific™
μPAC™ Neo Low-Load Trapping Columns
Combine μPAC™ Neo trapping columns with μPAC™ Neo low-load HPLC columns for optimal performance of bottom-up proteomics experiments.
Have Questions?
Catalog number COL-TRPLOLONEOB2
Price (USD)/ Pack of 2
950.00
Add to cart
Price (USD)/ Pack of 2
950.00
Add to cart
μPAC™ Neo Low-Load Trapping Columns
Catalog numberCOL-TRPLOLONEOB2
Price (USD)/ Pack of 2
950.00
-
Add to cart
Combine Thermo Scientific™ μPAC™ Neo low-load trapping columns with μPAC Neo low-load HPLC columns for optimal performance of bottom-up proteomics experiments. Specifically designed for single-cell proteomics applications, these trapping columns support the use of TMT-labeled samples, with up 10 ng of sample load.
Sample Preconcentration - Perform sample preconcentration in your preferred workflow, with both back flush and forward flush mobile phase direction capabilities.
Decreased Sample Loading Times - Shorten low-flow HPLC analysis run times by injecting your samples at 5-10 μL/min.
Sample Clean-up - Effectively remove salts, reagents, and contaminants that have no affinity for hydrophobic stationary phases from your samples, increasing your analytical column's lifetime and improving spectral quality and detection sensitivity.
Specifications
Flow Rate5 to 20 μL/min
Max. Pressure5800 psi (400 bar)
pH1.5 to 7
Porosity59%
Temperature40°C
Stationary PhaseC8
Packing MaterialSilicon Chip
Column TypeMicro Pillar Array
EndcappedYes
PhaseReversed Phase
Bed Length10 mm
Bed Width2 mm
Interpillar Distance2.5 μm
Pillar Diameter5 μm
Pillar Length18 μm
Unit SizePack of 2