COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters
Thermo Scientific™
COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters
Use COD (Chemical Oxygen Demand) Calibration Standards for Orion™ AQUAfast Colorimeters. Are ideal for environmental and wastewater monitoring.
Have Questions?
Catalog number CODS01
Price (USD)/ Each
74.67
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.40
Save 16.73 (18%)
-
Add to cart
Description:
AQUAfast 1000 ppm COD calibration standard, 475mL
Price (USD)/ Each
74.67
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.40
Save 16.73 (18%)
Add to cart
COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters
Catalog numberCODS01
Price (USD)/ Each
74.67
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.40
Save 16.73 (18%)
-
Add to cart
COD (Chemical Oxygen Demand) Calibration Standards are for use with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter.COD Chemistries can be measured by these colorimeters and ideal for environmental and wastewater monitoring. The calibration standards are used to check operations prior to using the instruments.
For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ2040
Two Calibration Standards: 1000 ppm and 10000 ppm, both in 475 mL bottles
Specifications
TypeCalibration Standards
For Use With (Equipment)AQ4000 Advanced Colorimeter, Orion AQUAfast II AQ2040 Colorimeter