Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent
Upgrade now to TrueCut Cas9 Protein v2 for the highest on-target editing efficiency.
Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent
Invitrogen™

Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is the first optimized lipid nanoparticle transfection reagent for CRISPR-Cas9 protein delivery. It is aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CMAX000080.75 mL
CMAX000030.3 mL
CMAX000010.1 mL
CMAX000151.5 mL
Catalog number CMAX00008
Price (USD)/ 0.75 mL
556.80
Special Offer
Online exclusive
Ends: 29-Nov-2024
640.00 
Save 83.20 (13%)
-
Add to cart
Quantity:
0.75 mL
Price (USD)/ 0.75 mL
556.80
Special Offer
Online exclusive
Ends: 29-Nov-2024
640.00 
Save 83.20 (13%)
Add to cart
Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent
Catalog numberCMAX00008
Price (USD)/ 0.75 mL
556.80
Special Offer
Online exclusive
Ends: 29-Nov-2024
640.00 
Save 83.20 (13%)
-
Add to cart
Lipofectamine CRISPRMAX Cas9 Transfection Reagent is the first optimized lipid nanoparticle transfection reagent for CRISPR-Cas9 protein delivery. It is a high-throughput-friendly, cost-effective alternative to electroporation.

Working together with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it provides:
• Demonstrated cleavage efficiency—tested in over 20 cell types including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and several others
• Low cell toxicity—less cells needed to initiate your experiment
• High throughput friendly—an ideal delivery solution for 96-well format
• Cost savings—whether cost-per-reaction or initial investment

Achieve high transfection efficiency in novel genome editing applications
Lipofectamine CRISPRMAX transfection reagent increases the likelihood of successful cleavage and recombination, especially when combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA. This ultimately maximizes the efficiency of performing genetic modifications.

Improve experimental outcomes using TrueCut Cas9 Protein v2
Unlike CRISPR plasmid or Cas9 mRNA, using Cas9 protein provides superior cleavage efficiency in primary cells and stem cells. It eliminates the need for transcription or translation of the payload, removes the risk of genomic integration, and is cell cycle independent.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free
Cell TypeEstablished Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample TypeProtein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection TechniqueLipid-based Transfection
For Use With (Application)Transfection
Product LineCRISPRMAX™, Lipofectamine™
Product TypeTransfection Reagent
Quantity0.75 mL
Serum CompatibleYes
Shipping ConditionApproved for shipment on Wet Ice
Unit Size0.75 mL
Contents & Storage
Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.