Gibco™ Hepatocyte Thaw Medium is a ready-to-use medium specifically designed for use with thawed cryopreserved hepatocytes to enhance their viabilityRead more
Have Questions?
Catalog number CM7500
Price (USD)/ 45 mL
94.65
Online exclusive
98.25
Save 3.60 (4%)
-
Add to cart
Price (USD)/ 45 mL
94.65
Online exclusive
98.25
Save 3.60 (4%)
Add to cart
Hepatocyte Thaw Medium
Catalog numberCM7500
Price (USD)/ 45 mL
94.65
Online exclusive
98.25
Save 3.60 (4%)
-
Add to cart
Gibco™ Hepatocyte Thaw Medium is a ready-to-use medium specifically designed for use with thawed cryopreserved hepatocytes to enhance their viability and overall quality. This medium is provided in a 50 mL conical tube for ease of use.
Features of Hepatocyte Thaw Medium:
• Easy to use—simply warm up to 37°C and add thawed hepatocytes • Recover cryopreserved hepatocytes with a single centrifugation • Designed for use with Gibco™ human and animal cryopreserved hepatocytes for optimal results • Provides high quality hepatocytes suitable for many in vitro assays, including induction, metabolic stability, and transporter assessments
Hepatocyte Thaw Medium is easy to use. Simply warm the tube of media to 37°C prior to thawing the cryopreserved hepatocytes. Once the medium is warm, thaw the cryopreserved hepatocytes per the Gibco™ protocol. Once the hepatocytes are thawed, pour the cells directly into the pre-warmed tube of Hepatocyte Thaw Medium and centrifuge. Remove the supernatant and proceed with the next steps of your experiment. There is no need to centrifuge the hepatocytes a second time.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHepatocyte Cells
ClassificationAnimal Origin
FormLiquid
Product TypeCell Culture Supplement
Serum LevelStandard Serum
With AdditivesPhenol Red
Format50 mL conical
Quantity45 mL
SourceAnimal origin, Human
SpeciesCattle/Bovine
Unit Size45 mL
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.