Catalog Number | Description |
---|---|
CHEM-CF-1 | NanoDrop CF-1 Calibration Check Fluid |
CHEM-CF-8 | CF-8 Calibration Check Kit |
CHEM-PR1-KIT | NanoDrop PR-1 Reconditioning Kit |
CHEM-PV-1 | NanoDrop PV-1 Performance Verification Solution |
CHEM-PV-8 | NanoDrop PV-8 Performance Verification Kit |
CHEM-PV-1 - PV-1 Performance Verification Solution
For use with NanoDrop One/OneC and NanoDrop Lite Plus
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
CHEM-PV-8 - PV-8 Performance Verification Kit
For use with NanoDrop Eight
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
CHEM-CF-1 - CF-1 Calibration Fluid
For use with NanoDrop 1000, NanoDrop 2000/2000c, NanoDrop Lite
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
CHEM-CF-8 - CF-8 Calibration Check Kit
For use with NanoDrop 8000
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
CHEM-PR1-KIT - PR-1 Reconditioning Kit
For use with all NanoDrop models
This kit is useful for reconditioning the sample measurement pedestals to a hydrophobic state if they become “unconditioned” (see user manual for more information). The PR-1 kit consists of a small screw cap vial of specially formulated compound and a supply of 25 convenient applicators.