The CyQUANT™ NF Cell Proliferation Assay provides a fast and sensitive method for counting cells in a population and measuringRead more
Have Questions?
Change view
Catalog Number
Quantity
C35007
1 Kit(s)
C35006
1000 reactions
2 Options
Catalog number C35007
Price (USD)/ 200 assays
193.65
Online exclusive
197.00
Save 3.35 (2%)
-
Add to cart
Quantity:
1 Kit(s)
Price (USD)/ 200 assays
193.65
Online exclusive
197.00
Save 3.35 (2%)
Add to cart
CyQUANT™ NF Cell Proliferation Assay
Catalog numberC35007
Price (USD)/ 200 assays
193.65
Online exclusive
197.00
Save 3.35 (2%)
-
Add to cart
The CyQUANT™ NF Cell Proliferation Assay provides a fast and sensitive method for counting cells in a population and measuring proliferation in microplate format.
Features of the CyQUANT™ NF Cell Proliferation Assay:
• More sensitive than MTT or AlamarBlue™ assays • Linear detection range from 100 to 20,000 cells per well (96-well microplate) • Assays can be completed in one hour
A Rapid and simple method for measuring cell proliferation The CyQUANT™ NF Cell Proliferation Assay does not require cell lysis, long incubations, radioactivity, or removal of stain from cells. The CyQUANT™ NF assay eliminates the freeze-thaw cell lysis step of the original CyQUANT™ cell proliferation assay by using a cell-permeant DNA-binding dye in combination with a plasma membrane permeabilization reagent. The CyQUANT™ NF Cell Proliferation Assay can be used in either 96-well or 384-well microplate formats, and is available in two configurations: a 200 assay kit (C35007) for smaller sample sizes, and a 1000 assay kit (C35006) for high-throughput applications.
AlamarBlue™ is a registered trademark of TREK Diagnostic Systems.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
For Use With (Application)Proliferation Assay
For Use With (Equipment)Microplate Reader
Product TypeCell Proliferation Assay
Dye TypeOther Label(s) or Dye(s)
Format384-well plate, 96-well plate
Product LineCyQUANT™
Quantity1 Kit(s)
Shipping ConditionRoom Temperature
Unit Size200 assays
Contents & Storage
Store in refrigerator 2°C to 8°C and protect from light.