Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit
Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit
Invitrogen™

Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit

Green features
The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit provides a fast, sensitive, non-toxic, and non-radioactive method for theRead more
Have Questions?
Catalog number C10428
Price (USD)/ 1 kit
782.65
Online exclusive
806.00 
Save 23.35 (3%)
-
Add to cart
Price (USD)/ 1 kit
782.65
Online exclusive
806.00 
Save 23.35 (3%)
Add to cart
Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit
Catalog numberC10428
Price (USD)/ 1 kit
782.65
Online exclusive
806.00 
Save 23.35 (3%)
-
Add to cart
The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit provides a fast, sensitive, non-toxic, and non-radioactive method for the detection of nascent protein synthesis utilizing fluorescence microscopy, high-content imaging, or flow cytometry. Included in the kit are L-homopropargylglycine (HPG), an amino acid analog of methionine containing an alkyne moiety, and Alexa Fluor™ 488 azide. The HPG is fed to cultured cells and incorporated into proteins during active protein synthesis. Addition of the Alexa Fluor™ 488 azide leads to a chemoselective ligation or 'click' reaction between the green fluorescent azide and the alkyne, allowing the modified proteins to be detected by imaged-based analysis.

Non-radioactive alternative—an alternative to the traditional 35S-methionine
Visualize bulk protein dynamics—fluorescent tagging of proteins allows their localization to be determined, including aggregation
Specificity—selective, specific reaction between label and detection tags
Stability —product contains an irreversible, covalent bond
Multiplex-enabled—use in conjuction with Click™-iT AHA (azide amino acid and alkyne dye) to detect spatial and temporal differences
Applicability to biological samples—easy detection; high sensitivity and low background, regardless of complexity

The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit has been successfully tested in HeLa, A549, and U-2 OS cells with a variety of reagents that inhibit protein synthesis, including cycloheximide and anisomycin. The applicability of these probes to monitor protein degradation has also been shown using inhibitors of the proteasome (MG132 and Bortezomib) and blockers of autophagy (chloroquine) in HeLa cells.

Additionally, due to differences in Click-iT™ chemistry between the Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit and Click™-iT AHA with Alexa Fluor™ 594 Alkyne, these reagents can be used in conjunction for spatial or temporal determination of differences in nascent protein synthesis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Detection MethodFluorescence
For Use With (Application)Protein Synthesis Assay
For Use With (Equipment)Fluorescence Microscope, Fluorescent Imager
Product TypeProtein Synthesis Assay Kit
Dye TypeProteins (Nascent)
EmissionVisible
FormLiquid
Green FeaturesLess hazardous
Product LineAlexa Fluor™, Click-iT™
Quantity1 kit
Unit Size1 kit
Contents & Storage
  • Store at 2°C to 6°C
  • DO NOT FREEZE
  • Protect material from long-term exposure to light, but may be exposed to light for short periods of time.