Human Dermal Fibroblasts, neonatal (HDFn)
Product Image
Gibco™

Human Dermal Fibroblasts, neonatal (HDFn)

Primary human dermal fibroblasts isolated from neonatal foreskin, cryopreserved at the end of the primary culture. • Viability at leastRead more
Have Questions?
Catalog number C0045C
Price (USD)/ 1 vial
660.65
Online exclusive
680.00 
Save 19.35 (3%)
-
Add to cart
Price (USD)/ 1 vial
660.65
Online exclusive
680.00 
Save 19.35 (3%)
Add to cart
Human Dermal Fibroblasts, neonatal (HDFn)
Catalog numberC0045C
Price (USD)/ 1 vial
660.65
Online exclusive
680.00 
Save 19.35 (3%)
-
Add to cart
Primary human dermal fibroblasts isolated from neonatal foreskin, cryopreserved at the end of the primary culture.
• Viability at least 70%
• ≥500,000 viable cells/vial
• Capable of at least 16 population doublings
• Mycoplasma: Not Detected
• Hepatitis B: Not Detected
• Hepatitis C: Not Detected
• HIV-1: Not Detected
• Bacteria, yeast, and other fungi: Not Detected
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AgeNewborn (14 days or less)
Cell TypeFibroblast Cells (Dermal)
Culture TypeAdherent Cell Culture
FormCryopreserved
SpeciesHuman
Donor SourceSingle Donor
No. of Cells≥500000
Product LineCascade Biologics™
Product TypeHuman Dermal Fibroblasts, Neonatal
Quantity1 vial
Tested ForHepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
Unit Size1 vial
Contents & Storage
Liquid nitrogen temperature