Bolt™ Transfer Buffer (20X)
Bolt™ Transfer Buffer (20X)
Invitrogen™

Bolt™ Transfer Buffer (20X)

Bolt Transfer Buffer (20X) is used to transfer proteins from Bolt Bis-Tris Plus gels to membranes for western blotting. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
BT000611L
BT0006125mL
Catalog number BT00061
Price (USD)/ 1 L
203.65
Online exclusive
218.00 
Save 14.35 (7%)
-
Add to cart
Quantity:
1L
Price (USD)/ 1 L
203.65
Online exclusive
218.00 
Save 14.35 (7%)
Add to cart
Bolt™ Transfer Buffer (20X)
Catalog numberBT00061
Price (USD)/ 1 L
203.65
Online exclusive
218.00 
Save 14.35 (7%)
-
Add to cart
Bolt Transfer Buffer (20X) is used to transfer proteins from Bolt Bis-Tris Plus gels to membranes for western blotting. It maintains the neutral pH environment established during electrophoresis. The neutral pH protects against modification of amino acid side chains and is compatible with N-terminal protein sequencing using Edman degradation.

Bolt Antioxidant may be used with Bolt Transfer Buffer to enhance transfer of reduced proteins to membranes.

See all available buffers and reagents available for SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferTransfer Buffers
Concentration20 X
Product TypeWestern Blot Transfer Buffer
Volume (Metric)1 L
FormLiquid
Gel CompatibilityBolt™ Bis-Tris Plus Gels
Product LineBolt™
Quantity1L
Unit Size1 L
Contents & Storage
Store at 4 to 25°C.