iCAP™ MTX Triple Quadrupole ICP-MS
iCAP™ MTX Triple Quadrupole ICP-MS
Thermo Scientific™
iCAP™ MTX Triple Quadrupole ICP-MS
Get advanced interference removal for your ICP-MS analysis with the Thermo Scientific™ iCAP™ MTX Triple Quadrupole ICP-MS system.
Have Questions?
Catalog number BRE731459A
Price (USD)/ Each
-
Request A Quote
Transform your research with the powerful interference-removal capabilities of the Thermo Scientific™ iCAP™ MTX Triple Quadrupole ICP-MS. This instrument reliably handles challenging samples with minimal complication, delivering right-first-time results alongside ease of use and continuity of operation. It offers greater confidence in your output as well as gains in productivity, giving you the time and expanded capabilities to unlock your analytical potential.
Advanced interference removal
  • Use of triple quadrupole for on-mass or mass-shift reactions
  • Reaction Finder for simple method development
  • Can be used as a single quadrupole for simple samples
Template-based workflow
  • Simple and intuitive
  • Fits into established laboratory practices
  • Ensures users are quickly up to speed
Optimized sample introduction
  • Unique peristaltic pump automatically tensioned for optimal performance
  • Maintains consistency
Auto-aligning torch
  • Quick-connect torch and injector system
  • Automatically aligned gas connections
  • No inconvenient manual adjustments required
Minimal cone cleaning
  • Advanced tuning
  • Minimal matrix deposits on cones
  • Limited cleaning requirements
  • Easy-access interface door for fast and convenient cone cleaning when required
User-friendly maintenance
  • Active instrument monitoring for consumables
  • Advanced scheduling of required maintenance
  • No user maintenance required downstream of the interface cones
  • Trouble-free operation
Specifications
TypeICP-MS (Triple Quadrupole )
DescriptioniCAP MTX Triple Quadrupole ICP-MS
Unit SizeEach