Future-proof your laboratory with the power of Triple Quadrupole ICP-MS. Exceptional interference removal for research level analysis combined with routine ease of use.
Delivering research level trace elemental analysis, combined with routine ease-of-use, the Thermo Scientific™ iCAP™ TQ ICP-MS is a high-performance, future-proof ICP-MS solution. Harness the power of Triple Quadropole (TQ) technology, in combination with a wide variety of reactive gases and experience, for uncomplicated analysis with incredible accuracy. Expand your applications and enhance your laboratory efficiency with breakthrough triple quadrupole technology that is so easy to use, any analyst can operate it.
Experience the benefits of research-level analysis and expand your laboratory's application portfolio, safe in the knowledge that your operators can seamlessly switch from our Single Quadrupole-ICP-MS instruments to Triple Quadrupole with Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS).
Whether you want to explore new markets, push the boundaries of your research, or meet future legislation requirements, the iCAP TQ ICP-MS instrument is the solution to future-proofing your lab.
Research level trace elemental analysis