Catalog Number | Description | Resolution |
---|---|---|
BRE725540 | Orbitrap Exploris GC Mass Spectrometer, no MS/MS | Up to 30,000 at m/z 200 |
BRE725541 | Orbitrap Exploris GC Mass Spectrometer, no MS/MS | Up to 60,000 at m/z 200 |
BRE725542 | Orbitrap Exploris GC Mass Spectometer with MS/MS | Up to 30,000 at m/z 200 |
BRE725543 | Orbitrap Exploris GC Mass Spectometer with MS/MS | Up to 60,000 at m/z 200 |
Deliver accurate quantitative results and spend less time reviewing data in high throughput analytical testing
Be one step ahead of changing demands
Focus on results, not instrument setup