Catalog Number | Description | Mass Range |
---|---|---|
BRE725533 | Orbitrap Exploris 480 MS with EASY-IC (internal calibration) | 40 to 6000 m/z |
BRE725539 | Orbitrap Exploris 480 MS with BioPharma option (factory installed) | 40 to 8000 m/z |
Maximize high-throughput performance in proteomics and biopharma applications with the Thermo Scientific Orbitrap Exploris 480 mass spectrometer. Where next level exploration delivers quantifiable accuracy, and selectivity required to obtain richer insights. Empowering translational scientists by bringing performance and intelligence together, the switch between exploration and targeted quantitation has never been easier. Suddenly your work goes deeper and even the routine will become simple, certain and extraordinary.
Maximum data certainty
Robustness and reliability
Empowers more users
High resolution within a compact footprint
Orbitrap power now for isotope ratios