Specifications
SpeciesBRD pathogens
Antimicrobial AgentsVarious
FormatSensititre™ Plate
Manufacturing OriginUnited Kingdom
Product TypeSensititre Vet Plate
Target Organism ClassFastidious and Non-fastidious Organisms
Test TypeMicrobial Susceptibility
For Use With (Application)Sensititre™
No. per Pack10
Packaging TypeBox
Quantity10
Shelf Life2 years
Dimensions (L x W)190 H x 200 W x 200 D mm
Unit SizeEach