Tyramide Conjugates
Tyramide Conjugates
Invitrogen™

Tyramide Conjugates

Thermo Fisher Scientific offers a wide variety of bright Alexa Fluor™, Alexa Fluor™ Plus or Biotin XX tyramide conjugates. These tyramide conjugates are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateExcitation/Emission
B56131Alexa Fluor™ Plus 750750/790 nm
B40952Alexa Fluor™ 350347/442 nm
B40953Alexa Fluor™ 488495/519 nm
B40954Alexa Fluor™ 546556/573 nm
B40955Alexa Fluor™ 555555/565 nm
B40956Alexa Fluor™ 568579/604 nm
B40957Alexa Fluor™ 594591/617 nm
B40958Alexa Fluor™ 647650/668 nm
B40951Biotin-XX
Catalog number B56131
Price (USD)/ 150 slides
484.65
Online exclusive
520.00 
Save 35.35 (7%)
-
Add to cart
Conjugate:
Alexa Fluor™ Plus 750
Excitation/Emission:
750/790 nm
Price (USD)/ 150 slides
484.65
Online exclusive
520.00 
Save 35.35 (7%)
Add to cart
Tyramide Conjugates
Catalog numberB56131
Price (USD)/ 150 slides
484.65
Online exclusive
520.00 
Save 35.35 (7%)
-
Add to cart
Thermo Fisher Scientific offers a wide variety of bright Alexa Fluor™, Alexa Fluor™ Plus or Biotin XX tyramide conjugates. These tyramide conjugates are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD). TSA is used to amplify signals up to 200 fold versus standard imaging methods. This reagent can be used with any antibody conjugated to HRP for tyramide signal amplification to detect low abundance targets. Up to seven-plex is possible in multiplexable fluorescent imaging.

These tyramide reagents are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD). TSA is used to amplify signal up to 200-fold versus standard imaging methods. These reagents are provided stand-alone, but are also available as part of our SuperBoost™ tyramide signal amplification kits (available separately). They can be used with any antibody conjugated to HRP for tyramide signal amplification for detection of low abundance targets. Up to seven-plex is possible in multiplex fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).

Features include:
• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with TO-PRO-3, secondary antibodies, and other SuperBoost kits
• Requires 10–100 times less primary antibody then standard ICC/IHC/ISH experiments

Tyramide reagents, when used as directed, can increase sensitivity up to 200 times over standard imaging methods. These reagents help sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal. They are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a tyramide reagent can be imaged using any type of microscope, producing high-resolution multiplex images.

Dissolve the Alexa Fluor™ or Biotin-XX tyramide reagent in 150 μL of DMSO to prepare 100X stock. Tyramide may coat the sides of the vial so invert the vial several times to ensure all reagent is dissolved in solution.

Specifications
ClonalityNone
Quantity1 Vial
For Use With (Application)Immunohistochemistry (IHC), Immunofluorescence (IF), Immunocytochemistry (ICC), ISH
Product LineAlexa Fluor™
ConjugateAlexa Fluor™ Plus 750
Excitation/Emission750/790 nm
Unit Size150 slides