Taq Buffers (10X)
Inquire about OEM or Commercial Supply version of this product here.
Product Image
Thermo Scientific™

Taq Buffers (10X)

Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
B33Taq Buffer with (NH4)2SO4
B55Taq Buffer without Detergent
B38Taq Buffer with KCl
B16Taq Buffer with KCl and MgCl2
B34Taq Buffer with (NH4)2SO4 and MgCl2
Catalog number B33
Price (USD)/ 4 x 1.25 mL
25.79
-
Add to cart
Product Type:
Taq Buffer with (NH4)2SO4
Request bulk or custom format
Price (USD)/ 4 x 1.25 mL
25.79
Add to cart
Taq Buffers (10X)
Catalog numberB33
Price (USD)/ 4 x 1.25 mL
25.79
-
Add to cart
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA polymerases (both recombinant and native). They are available in different compositions and also without detergents.

10X Taq Buffer without Detergent

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl

10X Taq Buffer with KCl and 15 mM MgCl2

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • 15 mM MgCl2
  • Detergent

10X Taq Buffer with (NH4)2SO4

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20

10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20
  • 20 mM MgCl2

10X Taq Buffer with KCl

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • Detergent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLiquid
PolymeraseTaq DNA Polymerase
Product TypeTaq Buffer with (NH4)2SO4
Quantity4 x 1.25 mL
Research CategoryPCR
Shipping ConditionWet or Dry Ice
For Use With (Application)Standard PCR
Concentration10X
No. of Reactions4 x 1.25 mL
Unit Size4 x 1.25 mL
Contents & Storage
• 10X Taq Buffer with (NH4)2SO4, 4 x 1.25 mL

Store at -20°C.