Perform differential pH measurements with a precalibrated plug-and-play sensor using the Thermo Scientific™ AquaSensors™ DataStick™ pH Measurement System. A replaceable reference junction contributes to long sensor life. Direct 24-bit data reporting is done via a variety of direct-to-PLC industrial communication protocols.