Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Actual product may vary
Thermo Scientific™

Orion™ AquaMate Vis and UV-Vis Spectrophotometers

Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSpectral Bandwidth
AQ7100AquaMate 7100 Vis Spectrophotometer with North America, European and UK Power cord5 nm
AQ7100APACAquaMate 7100 Vis Spectrophotometer with China, Indian and Australia / NZ Power cord5 nm
AQ8100AquaMate 8100 UV-Vis Spectrophotometer with North America, European and UK Power cord2 nm
AQ8100APACAquaMate 8100 Vis Spectrophotometer with China, Indian and Australia / NZ Power cord2 nm
Catalog number AQ7100
Price (USD)/ Each
4,608.45
Special Offer
Online exclusive
Ends: 31-Dec-2024
5390.00 
Save 781.55 (15%)
-
Add to cart
Description:
AquaMate 7100 Vis Spectrophotometer with North America, European and UK Power cord
Spectral Bandwidth:
5 nm
Price (USD)/ Each
4,608.45
Special Offer
Online exclusive
Ends: 31-Dec-2024
5390.00 
Save 781.55 (15%)
Add to cart
Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Catalog numberAQ7100
Price (USD)/ Each
4,608.45
Special Offer
Online exclusive
Ends: 31-Dec-2024
5390.00 
Save 781.55 (15%)
-
Add to cart
Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.

Pre-programmed Methods:

  • More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Glove friendly 7-inch touch screen controller
  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further
Specifications
Baseline Flatness±0.005 A
Certifications/ComplianceCE / TUV
CompartmentsAccessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down
ConnectionsSingle USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Depth (Metric)35.5 cm
DescriptionAquaMate 7100 Vis Spectrophotometer with North America, European and UK Power cord
Detector TypeDual Silicon Photodiodes
Display7 in. High Definition Color Touchscreen, Fixed, 800 x 1280 pixels
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Light SourceTungsten Halogen
Spectral Bandwidth5 nm
TypeVisible Specrtophotometer
Wavelength Range325 to 1100 nm
Drift<0.0010 A/hr. (At 500 nm after 1 hour warmup)
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
For Use With (Application)Water and Waste Water Analysis
Height (Metric)19.5 cm
Includes
Test Tube Holder for vials and test tube from 12 to 25 mm OD, AquaMate Water Analysis Methods Software, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362).
InterfaceGlove friendly touch-screen
KeypadGlove- friendly touchscreen
Lamp Life>1000 hrs.
Length (Metric)35.5 cm
Noise(RMS at 500 nm 60 consecutive measurements) ≤0.00020A at 0A at 260 and 500 nm ≤0.00030A at 1A at 260 and 500 nm ≤0.00040A at 2A at 260 and 500 nm
Optical DesignDouble Beam
Photometric Accuracy±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A
Photometric Display3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Printer IncludedSnap-on printer available
Scan SpeedAutomatic – up to 1800 nm/min.
Stray Light<0.05%T at 340 nm and 400 nm
Warranty1 Year
Wavelength Accuracy±0.5 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Weight (Metric)7.5 kg
Width (Metric)38.5 cm
Unit SizeEach