Orion™ AquaMate Preprogrammed Methods on USB Memory Stick
Orion™ AquaMate Preprogrammed Methods on USB Memory Stick
Thermo Scientific™
Orion™ AquaMate Preprogrammed Methods on USB Memory Stick
Use the Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick with over 260 preprogrammed methods on the Orion™ AquaMate spectrophotometers
Have Questions?
Catalog number AQ0USB
Price (USD)/ Each
61.30
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.70 
Save 10.40 (15%)
-
Add to cart
Type:
USB Memory Stick
Price (USD)/ Each
61.30
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.70 
Save 10.40 (15%)
Add to cart
Orion™ AquaMate Preprogrammed Methods on USB Memory Stick
Catalog numberAQ0USB
Price (USD)/ Each
61.30
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.70 
Save 10.40 (15%)
-
Add to cart
Use the Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick with over 260 preprogrammed methods for Orion™, CHEMetrics and Merck reagent chemistries on the Orion™ AquaMate 8000 UV-Vis and Orion™ AquaMate 7000 Vis spectrophotometers. Preprogrammed methods can be added, removed or modified on the Orion™ AquaMate spectrophotometers at any time.
  • Compatible with the AquaMate 8000 UV-Vis and AquaMate 7000 Vis Spectrophotometers.
Specifications
TypeUSB Memory Stick
For Use With (Equipment)Orion AquaMate spectrophotometers
IncludesOver 260 preprogrammed methods compatible with several leading brands of reagents
Unit SizeEach