EVOS™ Onstage Vessel Holder, Two 35 mm Petri Dish
EVOS™ Onstage Vessel Holder, Two 35 mm Petri Dish
EVOS™ Onstage Vessel Holder, Two 35 mm Petri Dish
EVOS Onstage vessel holders maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FLRead more
Have Questions?
Catalog number AMEPVH030
Price (USD)/ 1 each
385.00
-
Add to cart
Price (USD)/ 1 each
385.00
Add to cart
EVOS™ Onstage Vessel Holder, Two 35 mm Petri Dish
Catalog numberAMEPVH030
Price (USD)/ 1 each
385.00
-
Add to cart
EVOS Onstage vessel holders maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with an EVOS Onstage Incubator. EVOS Onstage vessel holders allow a perfect fit of your micro-well plates or cell culture flasks/dishes to the master plate of the EVOS Onstage Incubator for increased precision in sample alignment. This particular vessel holder accommodates two 35-mm Petri dishes. With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy.
Learn more about the EVOS FL Auto Imaging System.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeVessel Holder
Product LineEVOS
For Use With (Equipment)EVOS™ FL Auto Imaging System
HoldsHolds two 35 mm Petri dishes
Quantity1 each
Unit Size1 each