EVOS™ Vessel Holder, Nunc™/SPL IVF 4-well dish
EVOS™ Vessel Holder, Nunc™/SPL IVF 4-well dish
EVOS™ Vessel Holder, Nunc™/SPL IVF 4-well dish
EVOS vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate toRead more
Have Questions?
Catalog number AMEPVH011
Price (USD)/ 1 each
129.00
-
Add to cart
Price (USD)/ 1 each
129.00
Add to cart
EVOS™ Vessel Holder, Nunc™/SPL IVF 4-well dish
Catalog numberAMEPVH011
Price (USD)/ 1 each
129.00
-
Add to cart
EVOS vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment. This particular vessel holder accommodates one Nunc or SPL IVF 4-well dish. It is just one of many easy-to-change vessel holders and stage plates available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Note: an adapter is needed for use of this holder on the EVOS FL Auto automated stage.
For additional choices, see the EVOS Guide to Vessel Holders and Stage Plates.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeVessel Holder
Product LineEVOS
For Use With (Equipment)EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
FormatNunc 4-well dish for IVF
Holds1 Nunc/SPL IVF 4-well Dish
Quantity1 Holder
Unit Size1 each