This Celleste 3D Analysis Module, single-user license, helps visualize complex 3D objects using guided segmentation designed to facilitate identification and measurements. This module will enable users to threshold and generate iso-surfaces as well as measure distances, volumes, and areas, and perform other measurements. This module is used in conjunction with
Celleste 6.0 Image Analysis Software.
Celleste 6 software is a suite of image analysis and visualization tools designed for a range of biological applications that lets you transform your images into quantitative data in a streamlined and customizable workflow. Coupled with the powerful image acquisition capabilities of our
EVOS microscopes, Celleste 6 software allows you to seamlessly capture, process, measure, analyze, and share images and data. The 3D analysis module requires the use of the
Celleste 3D Visualization Module, single-user license.
Celleste 6.0 software with the 3D Analysis Module offers you these important advantages:
• Icon-based wizard-driven workflow streamlines image analysis and eliminates guesswork• Preconfigured and optimized analysis templates for common applications help derive the most relevant results from image sets
• Wide range of report formats to visualize results, create reports, and share images and data
• Automatic segmentation of objects using any 2D frame in the Z-stack
• Seamless propagation t in Z with automated or semi-automated tools
• Automatic generation of iso-surfaces and pseudo-surfaces
• 3D counting, including co-localization
• Measurement of point-to-point distances, surfaces, and volumes
Celleste 6 software is a powerful image analysis solution that helps make your imaging research job easier and increases your productivity by seamlessly providing images and accurate data. Coupled with the powerful image acquisition capabilities of the EVOS imaging system, you have the tools that make it easy to capture, process, measure, analyze, and share your important images and data more effectively than ever.
For Research Use Only. Not for use in diagnostic procedures.