TE, pH 7.0, RNase-free
Inquire about OEM or Commercial Supply version of this product here.
TE, pH 7.0, RNase-free
Invitrogen™
TE, pH 7.0, RNase-free
Ambion TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM986150 mL
AM986010 x 1 mL
Catalog number AM9861
Price (USD)/ 50 mL
58.65
Online exclusive
60.75 
Save 2.10 (3%)
-
Add to cart
Quantity:
50 mL
Price (USD)/ 50 mL
58.65
Online exclusive
60.75 
Save 2.10 (3%)
Add to cart
TE, pH 7.0, RNase-free
Catalog numberAM9861
Price (USD)/ 50 mL
58.65
Online exclusive
60.75 
Save 2.10 (3%)
-
Add to cart
Ambion TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water. It is provided in one bottle containing 50 mL. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. TE Buffer is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore at room temperature.
pH7.0
Physical FormLiquid
Product LineAmbion
Quantity50 mL
PackagingBottle
Unit Size50 mL