Formamide (Deionized)
Inquire about OEM or Commercial Supply version of this product here.
Product Image
Invitrogen™

Formamide (Deionized)

Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon.Read more
Have Questions?
Catalog number AM9344
Price (USD)/ 5 x 500 g
502.65
Online exclusive
538.00 
Save 35.35 (7%)
-
Add to cart
Quantity:
5 x 500 g
Price (USD)/ 5 x 500 g
502.65
Online exclusive
538.00 
Save 35.35 (7%)
Add to cart
Formamide (Deionized)
Catalog numberAM9344
Price (USD)/ 5 x 500 g
502.65
Online exclusive
538.00 
Save 35.35 (7%)
-
Add to cart

Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon. Supplied in 5 bottles containing 500 g each. Formamide lowers the melting temperature of nucleic acid duplexes and is widely used in applications such as Southern and northern blotting, nuclease protection assays, S1 mapping, and in situ hybridization. For critical applications, it is recommended that the formamide from previously opened bottles be deionized before use.

Appearance: Clear, colorless
Molecular Weight: 45.04
Purity: ≥99.5%
Quality Control: The product has been tested for RNase and DNase contamination.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorColorless
Packaging TypeBottle
Product TypeFormamide
Product LineAmbion™
Purity>99.5%
Quantity5 x 500 g
FormLiquid
Unit Size5 x 500 g
Contents & Storage
Store at 4° or -20°C as small aliquots.