New RNAi Support Center—Find technical tips and troubleshooting recommendations for your RNAI experiments from our scientists.
Invitrogen™
Pre-miR™ miRNA Precursor Negative Control #1
Pre-miR™ miRNA Precursors are designed to mimic endogenous miRNAs. Pre-miR miRNA Precursor Negative Control #1 is designed to use asRead more
Have Questions?
Catalog number AM17110
Price (USD)/ 5 nmol
337.65
Online exclusive
361.00
Save 23.35 (6%)
-
Add to cart
Price (USD)/ 5 nmol
337.65
Online exclusive
361.00
Save 23.35 (6%)
Add to cart
Pre-miR™ miRNA Precursor Negative Control #1
Catalog numberAM17110
Price (USD)/ 5 nmol
337.65
Online exclusive
361.00
Save 23.35 (6%)
-
Add to cart
Pre-miR™ miRNA Precursors are designed to mimic endogenous miRNAs. Pre-miR miRNA Precursor Negative Control #1 is designed to use as a negative control for experiments using Pre-miR miRNA Precursors. Transfect Pre-miR miRNA Precursor Negative Control #1 using the same methodology as for your experimental Pre-miR miRNA Precursors, then use the target gene expression from the sample transfected with the Pre-miR miRNA Precursor Negative Control #1 as a baseline for evaluation of the effect of the experimental Pre-miR miRNA Precursor on target gene expression. Advantages of the Pre-miR™ miRNA Precursor Negative Control #1:
• Optimized design results in correct strand utilization by RISC • Purified and ready to transfect
About Pre-miR miRNA Precursors Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments.
Using Pre-miR miRNA Precursor Negative Control #1 Pre-miR™ Negative Control #1 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. RNA oligonucleotides are susceptible to degradation by exogenous ribonucleases introduced during handling. Wear gloves when handling this product. Use RNase-free reagents, tubes, and barrier pipette tips. Upon receipt, store in a non-frost-free freezer at or below -20°C (dried oligonucleotides are shipped at ambient temperature).
NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Control TypeNegative Control
RNAi TypemiRNA Mimic
For Use With (Application)miRNA & Non-Coding RNA Analysis
Label or DyeUnconjugated
Product LineAmbion™, Pre-miR™
Product TypemiRNA Mimic
Quantity5 nmol
Shipping ConditionRoom Temperature
Unit Size5 nmol
Contents & Storage
Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.