Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Actual product may vary
Thermo Scientific™

Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents

Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionRange
ACR012AQUAfast ammonia LR reaction tubes, 50 tubes0.02 to 2.5mg/L N
AC2009AQUAfast fluoride SPADNS liquid reagent, 50 tests0.05 to 2mg/L F
AC3001AQUAfast pH liquid dropper reagent, 30 tests6.5 to 8.4 pH
AC4P54AQUAfast manganese LR reagent powder & liquid set, 50 tests0.01 to 0.7mg/L Mn
ACD004AQUAfast total nitrogen LR digestion tubes, 50 pack0.5 to 25mg/L N
ACD007AQUAfast total nitrogen HR digestion tubes, 50 pack5 to 150mg/L N
ACD095AQUAfast phosphate (total) digestion tubes, 50 pack0.02 to 1.1mg/L P
ACD095AHAQUAfast phosphate (acid hydrolysable) digestion tubes, 50 pack0.02 to 1.6mg/L P
ACR007AQUAfast nitrate reaction tubes, 50 pack1 to 30mg/L N
ACR011AQUAfast ammonia HR reaction tubes, 50 tubes1 to 50mg/L N
ACR095AQUAfast phosphate (ortho) reaction tubes, 50 pack0.06 to 5mg/L PO4
CODH00AQUAfast COD 0-1500 ppm digestion tubes, 25 pack0 to 1500mg/L O2
CODHP0AQUAfast COD 0-15000 ppm digestion tubes, 25 pack0 to 15000mg/L O2
CODL00AQUAfast COD 0-150 ppm digestion tubes, 25 pack0 to 150mg/L O2
CODH150AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range0 to 15000 mg/L O2
CODL150AQUAfast COD 0-150 ppm digestion tubes, 150 pack0 to 150 mg/L O2
CODM150AQUAfast COD 0-1500 ppm digestion tubes, 150 pack0 to 1500 mg/L O2
Catalog number ACR012
Price (USD)/ Each
110.30
Special Offer
Online exclusive
Ends: 31-Dec-2024
129.00 
Save 18.70 (15%)
-
Add to cart
Description:
AQUAfast ammonia LR reaction tubes, 50 tubes
Range:
0.02 to 2.5mg/L N
Price (USD)/ Each
110.30
Special Offer
Online exclusive
Ends: 31-Dec-2024
129.00 
Save 18.70 (15%)
Add to cart
Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Catalog numberACR012
Price (USD)/ Each
110.30
Special Offer
Online exclusive
Ends: 31-Dec-2024
129.00 
Save 18.70 (15%)
-
Add to cart
Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.
Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.

 

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Digestion tube tests and reaction tube tests are preprogrammed in the Orion AQ3700 meter. Some of the listed tests have only liquid components, like our AC2009 SPADNS test for fluoride, and some have liquid and powder or liquid and tablet reagents.
Specifications
DescriptionAQUAfast ammonia LR reaction tubes, 50 tubes
EPA Approved MethodsWastewater: Standard Methods 4500-NH3 F-1997
FormatReaction Tube
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter
IncludesSet of 3: 50 Ammonia Diluent Reagent LR Reaction Tubes, 50 Ammonia Salicylate Powder Packs, 50 Ammonia Cyanurate Powder Packs
Quantity50 Tubes
Range0.02 to 2.5mg/L N
Test MethodSalicylate
TypeAmmonia
Unit SizeEach