Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Actual product may vary
Thermo Scientific™
Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Ensure your turbidity meter has the accessories it needs, including turbidity vials, primary turbidity standards, Formazin turbidity standards, and more.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
AC45S1Silicon Oil and Cloth Kit
AC45FZFormazin turbidity standard, 4000NTU, 475mL
AC45STPrimary Standards for AQ4500, SDVB for 0, 1.0, 10, 100, 1000NTU
AC2T24Replacement Turbidity Vials
AQ4CBLRS232 cable for method uploads and data downloads in AQ4000/AQ4500
Catalog number AC45S1
Price (USD)/ Each
29.16
Special Offer
Online exclusive
Ends: 31-Dec-2024
34.10 
Save 4.94 (14%)
-
Add to cart
Description:
Silicon Oil and Cloth Kit
Price (USD)/ Each
29.16
Special Offer
Online exclusive
Ends: 31-Dec-2024
34.10 
Save 4.94 (14%)
Add to cart
Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Catalog numberAC45S1
Price (USD)/ Each
29.16
Special Offer
Online exclusive
Ends: 31-Dec-2024
34.10 
Save 4.94 (14%)
-
Add to cart
Ensure your Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter has the accessories it needs with our selection of Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories that include turbidity vials, primary turbidity standards, Formazin turbidity standards, and more. Our Orion AQUAfast AQ4500 turbidity meter has advanced features, including dual light sources and drinking water / wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols; use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
Accessories for the Orion AQUAfast turbidity meter include primary standards, Formazin turbidity standard, silicon oil and cloth kit, and replacement turbidity vials.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeSilicon Oil and Cloth Kit
For Use With (Equipment)AQ4500 Turbidimeter
DescriptionSilicon Oil and Cloth Kit
EPA Approved MethodsUS EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7028
Unit SizeEach