Orion™ AQUAfast™ IV Chemistry Reagents
Orion™ AQUAfast™ IV Chemistry Reagents
Actual product may vary
Thermo Scientific™
Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberRangeTypeMethod
AC40780.2 to 6 mg/L FeIronPhenanthroline
AC40040.2 to 1.5 mg/L NNitrateCadmium Reduction
AC40060.04 to 0.4 mg/LCyanide
AC40051.0 to 7.5 mg/L NNitrateCadmium Reduction
AC40075 to 50 mg/L NO3NitrateCadmium Reduction
AC40100.2 to 3 mg/L NAmmoniaSalicylate
AC40111.5 to 14 mg/L NAmmoniaDirect Nesslerization
AC40120.5 to 7 mg/L NAmmoniaDirect Nesslerization
AC40160.2 to 3 mg/L SSulfideMethylene Blue
AC40172.5 to 40 mg/L Cl-ChlorideFerric Thiocyanide
AC40270.04 to 0.25 mg/L AlAluminumEriochrome Cyanine R
AC40460.08 to 1 mg/L NNitriteDiazotization (Azo)
AC40480.2 to 5 mg/L O3OzoneDPD
AC40552 to 30 mg/L MnManganesePeriodate
AC40600.5 to 10 mg/L SiO2SilicaHeteropoly Blue
AC40650.3 to 3 mg/L ZnZincZincon
AC40700.4 to 5 mg/L Cl2ChlorineDPD
AC408210 to 100 mg/L SO4SulfateBariumsulfate-Turbidity
AC40950.3 to 8 mg/L PO4PhosphateStannous Chloride
AC40990.8 to 11 mg/L ClO2Chlorine DioxideDPD
Catalog number AC4078
Price (USD)/ Case of 30
64.53
Online exclusive
71.70 
Save 7.17 (10%)
-
Add to cart
Range:
0.2 to 6 mg/L Fe
Type:
Iron
Method:
Phenanthroline
Price (USD)/ Case of 30
64.53
Online exclusive
71.70 
Save 7.17 (10%)
Add to cart
Orion™ AQUAfast™ IV Chemistry Reagents
Catalog numberAC4078
Price (USD)/ Case of 30
64.53
Online exclusive
71.70 
Save 7.17 (10%)
-
Add to cart
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeIron
Test Reagent FormatsAmpoule
For Use With (Equipment)Orion AQUAfast AQ4000 Colorimeter
DescriptionAQUAfast iron (total & soluble) Auto-ID ampoules, 30 pack
Includes30 Auto-ID Ampoules
MethodPhenanthroline
For Use With (Application)Colorimetric water and wastewater testing equipment
Recommended AccessoriesAQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Range0.2 to 6 mg/L Fe
EPA Approved MethodsWastewater: Standard Methods 3500-Fe-1997
Quantity30 Ampoules
Unit SizeCase of 30