Catalog Number | Range | Type | Method |
---|---|---|---|
AC4006 | 0.04 to 0.4 mg/L | Cyanide | |
AC4004 | 0.2 to 1.5 mg/L N | Nitrate | Cadmium Reduction |
AC4005 | 1.0 to 7.5 mg/L N | Nitrate | Cadmium Reduction |
AC4007 | 5 to 50 mg/L NO3 | Nitrate | Cadmium Reduction |
AC4010 | 0.2 to 3 mg/L N | Ammonia | Salicylate |
AC4011 | 1.5 to 14 mg/L N | Ammonia | Direct Nesslerization |
AC4012 | 0.5 to 7 mg/L N | Ammonia | Direct Nesslerization |
AC4016 | 0.2 to 3 mg/L S | Sulfide | Methylene Blue |
AC4017 | 2.5 to 40 mg/L Cl- | Chloride | Ferric Thiocyanide |
AC4027 | 0.04 to 0.25 mg/L Al | Aluminum | Eriochrome Cyanine R |
AC4046 | 0.08 to 1 mg/L N | Nitrite | Diazotization (Azo) |
AC4048 | 0.2 to 5 mg/L O3 | Ozone | DPD |
AC4055 | 2 to 30 mg/L Mn | Manganese | Periodate |
AC4060 | 0.5 to 10 mg/L SiO2 | Silica | Heteropoly Blue |
AC4065 | 0.3 to 3 mg/L Zn | Zinc | Zincon |
AC4070 | 0.4 to 5 mg/L Cl2 | Chlorine | DPD |
AC4078 | 0.2 to 6 mg/L Fe | Iron | Phenanthroline |
AC4082 | 10 to 100 mg/L SO4 | Sulfate | Bariumsulfate-Turbidity |
AC4095 | 0.3 to 8 mg/L PO4 | Phosphate | Stannous Chloride |
AC4099 | 0.8 to 11 mg/L ClO2 | Chlorine Dioxide | DPD |
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.