Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Thermo Scientific™
Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
AC3V2525mm Vials, 3 Pack
AC3CBRVial Cleaning Brushes, 5 Pack
Catalog number AC3V25
Price (USD)/ Each
117.14
Special Offer
Online exclusive
Ends: 31-Dec-2024
137.00 
Save 19.86 (15%)
-
Add to cart
Description:
25mm Vials, 3 Pack
Price (USD)/ Each
117.14
Special Offer
Online exclusive
Ends: 31-Dec-2024
137.00 
Save 19.86 (15%)
Add to cart
Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Catalog numberAC3V25
Price (USD)/ Each
117.14
Special Offer
Online exclusive
Ends: 31-Dec-2024
137.00 
Save 19.86 (15%)
-
Add to cart
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Additional Accessory: 
  • Replacement Calibration Standards (see AC301S)
Specifications
For Use With (Equipment)AQUAfast AQ3010 Turbidity Meter
Description25mm Vials, 3 Pack
Unit SizeEach