Orion™ AQUAfast™ II Chemistries Test Kits
Orion™ AQUAfast™ II Chemistries Test Kits
Thermo Scientific™
Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
AC3072AQUAfast chlorine (total) HR & sodium chlorate reagent tablets, 100 tests
AC2002AQUAfast alkalinity-m reagent tablets, 100 tests
AC3002PAQUAfast alkalinity-p reagent tablets, CaCO3, 100 tests
AC2027AQUAfast aluminum reagent tablets, 50 tests
AC4P27AQUAfast Aluminum Powder Pack, 100 tests
AC2012AQUAfast ammonia reagent tablets, 50 tests
AC2035Bromine
AC2017AQUAfast chloride reagent tablets, 50 tests
AC2099AQUAfast chlorine dioxide reagent tablets, 100 tests
AC2071AQUAfast chlorine (free) reagent tablets, 100 tests
AC2070AQUAfast chlorine (free & total) reagent tablets, 50 tests each
AC2072AQUAfast chlorine (total) reagent tablets, 100 tests
AC2029AQUAfast copper (free & total) reagent tablets, 50 tests
AC2065AQUAfast zinc reagent tablets, 50 tests
AC2098AQUAfast cyanuric acid reagent tablets, 100 tests
AC3032C2AQUAfast hardness (calcium) LR reagent tablets, Murexide Method, 100 tests
AC3032CAQUAfast hardness (calcium) HR reagent tablets, Murexide Method, 100 tests
AC3032TAQUAfast hardness (total) reagent tablets, Metallphthalein Method, 100 tests
AC2030AQUAfast hydrazine reagent powder kit, 30 tests
AC3069AQUAfast hydrogen peroxide reagent tablets, 100 tests
AC2078AQUAfast iron (II & III) reagent tablets, 100 tests
AC2055AQUAfast manganese reagent tablets, 50 tests
AC2007AQUAfast nitrate reagent tablets, 50 tests
AC2046AQUAfast nitrite reagent tablets, 100 tests
AC2048AQUAfast ozone reagent tablets, 100 tests
AC2001AQUAfast pH reagent tablets, 100 tests
AC3001BPAQUAfast ph LR reagent tablets, bromocresol purple, 100 tests
AC3001TBAQUAfast pH HR reagent tablets, thymol blue, 100 tests
AC2095-WAAQUAfast phosphate (ortho) LR reagent tablets, 50 tests
AC2096AQUAfast phosphate (ortho) HR reagent tablets, 50 tests
AC3019AQUAfast potassium reagent tablets, 100 tests
AC2060AQUAfast silica reagent tablets, 50 tests
AC2061AQUAfast silica phosphate removal reagent, 100 tablets
AC2082AQUAfast sulfate reagent tablets, 100 tests
AC2016AQUAfast sulfide reagent tablets, 100 tests
Catalog number AC3072
Price (USD)/ Each
74.90
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.60 
Save 12.70 (15%)
-
Add to cart
Description:
AQUAfast chlorine (total) HR & sodium chlorate reagent tablets, 100 tests
Price (USD)/ Each
74.90
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.60 
Save 12.70 (15%)
Add to cart
Orion™ AQUAfast™ II Chemistries Test Kits
Catalog numberAC3072
Price (USD)/ Each
74.90
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.60 
Save 12.70 (15%)
-
Add to cart
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeChlorine, Sodium Chlorate
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter
DescriptionAQUAfast chlorine (total) HR & sodium chlorate reagent tablets, 100 tests
Range5 to 200 mg/L
Unit SizeEach