ABsolute Blue QPCR Mix, SYBR Green, ROX
ABsolute Blue QPCR Mix, SYBR Green, ROX
Thermo Scientific™

ABsolute Blue QPCR Mix, SYBR Green, ROX

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB4162B
also known as AB-4162/B
1600 Reactions
AB4163A
also known as AB-4163/A
400 Reactions
Catalog number AB4162B
also known as AB-4162/B
Price (USD)/ 1,600 reactions
2,975.00
-
Add to cart
No. of Reactions:
1600 Reactions
Price (USD)/ 1,600 reactions
2,975.00
Add to cart
ABsolute Blue QPCR Mix, SYBR Green, ROX
Catalog numberAB4162B
Price (USD)/ 1,600 reactions
2,975.00
-
Add to cart
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of ABsolute Blue QPCR Mix, SYBR Green, and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.

Highlights

High performance—ABsolute sensitivity, consistency & convenience
Enhanced contrast and reduced errors—easily identify correctly aliquotted reaction mix in all plates and tubes
Universal—compatible with all major qPCR platforms
Adaptable—available with ROX or Fluorescein passive reference dyes

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Includes
• ABsolute Blue qPCR SYBR Green ROX Mix contains 2X master mix supplemented with high ROX reference dye concentration

Related Products
ABsolute Blue QPCR Mix, low ROX
ABsolute Blue QPCR Mix, with separate ROX vial
ABsolute Blue QPCR Mix, ROX
ABsolute Blue QPCR Mix, SYBR Green, low ROX
ABsolute Blue QPCR Mix, SYBR Green, ROX
ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial
ABsolute Blue QPCR Mix, SYBR Green, fluorescein
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodSYBR
FormatTube
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
PolymeraseTaq DNA Polymerase
Reaction SpeedStandard
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product LineABsolute Blue
Product TypeReal Time PCR SYBR Master Mix
Sufficient For1600 Reactions of 25 μL Each
For Use With (Application)Gene Expression, Genotyping, siRNA Validation
CompatibilityqPCR Cyclers
Concentration2X
No. of Reactions1600 Reactions
Sample TypeDNA, dsDNA
Volume16 x 1.25 mL
Unit Size1,600 reactions
Contents & Storage
Contains:
• 16 x 1.25 mL ABsolute Blue QPCR SYBR Green ROX Mix (2X)
• 100 μL MgCl2 (1M)
Sufficient for 1600 x 25 μL reactions.

Store at -20°C