Specifications
BarcodeNo Barcode
BreakapartYes
Packaging25 Plates
Product LineArmadillo
ColorClear
Product TypePCR Microplate
For Use With (Equipment)Real-Time PCR Systems or Thermal Cycler
Skirt StyleSemi
MaterialPolycarbonate
Volume (Metric) Well0.2 mL
No. of Wells96
Volume (Metric) Working0.1 mL
Well ShapeRound Bottom (U-well)
Quantity25 plates
Reaction SpeedLow Profile
Well ColorWhite
Unit Size25 plates