ABsolute QPCR Mix, no ROX
Product Image
Thermo Scientific™

ABsolute QPCR Mix, no ROX

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB1133A400 Reactions
AB1132B
also known as AB-1132/B
1600 Reactions
Catalog number AB1133A
Price (USD)/ 400 reactions
790.00
-
Add to cart
No. of Reactions:
400 Reactions
Price (USD)/ 400 reactions
790.00
Add to cart
ABsolute QPCR Mix, no ROX
Catalog numberAB1133A
Price (USD)/ 400 reactions
790.00
-
Add to cart
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute QPCR Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes. The 2X master mix contains a proprietary reaction buffer that provides high sensitivity, specificity, and reproducibility. ABsolute qPCR Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, and with no ROX reference dye. There is also a master mix version optimized for capillary instruments.

Highlights

• High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
• Optimised buffer - For maximal performance in multiplex reactions

Applications
• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodPrimer-probe
PCR MethodqPCR
PolymeraseTaq DNA Polymerase
Reaction SpeedStandard
For Use With (Equipment)BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon
Product LineABsolute
Product TypeAbsolute QPCR Mix, No Rox
Sufficient For400 Reactions
No. of Reactions400 Reactions
Sample TypeDNA, dsDNA
Unit Size400 reactions
Contents & Storage
Contains:
• 1 x 5 mL ABsolute QPCR Mix (2X)

Store at -20°C