Ultra Clear qPCR Caps, strips of 8
Try our New Thermo Scientific PCR Plastics, Sustain Series - identical performance and quality with a lower carbon footprint
Ultra Clear qPCR Caps, strips of 8
Thermo Scientific™
Ultra Clear qPCR Caps, strips of 8
Thermo Scientific Ultra Clear qPCR Cap Strips of 8 have a highly polished flat surface, providing an optically clear windowRead more
Have Questions?
Catalog number AB0866
Price (USD)/ 120 strips
59.65
Online exclusive
66.75 
Save 7.10 (11%)
Add to cart
Price (USD)/ 120 strips
59.65
Online exclusive
66.75 
Save 7.10 (11%)
Add to cart
Ultra Clear qPCR Caps, strips of 8
Catalog numberAB0866
Price (USD)/ 120 strips
59.65
Online exclusive
66.75 
Save 7.10 (11%)
-
Add to cart
Thermo Scientific Ultra Clear qPCR Cap Strips of 8 have a highly polished flat surface, providing an optically clear window to deliver maximum and consistent signal transmission, critical for accurate qPCR results. The caps are molded from a specially formulated polypropylene that enhances signal detection by minimizing autofluorescense and maximizing transmission. The Ultra Clear qPCR Cap Strips are compatible with all Thermo Scientific PCR tube strips and 96-well PCR plates.
Features of Thermo Scientific Ultra Clear qPCR Cap Strips
• 8-cap strip
• Compatible with all Thermo Scientific 96-well PCR plates as well as standard and low profile tube strips
• Specially formulated ultra clear polypropylene
• Highly polished
• Flat cap design
Specifications
ColorClear
MaterialPolypropylene
No. of Wells8
Quantity120/Cs.
FormatStrips of 8 Optical Caps
Product TypePCR Plate Cap Strip
Certified RNase/DNase-freeYes
Plate Compatibility96-well Plate
Unit Size120 strips