Specifications
Certifications/ComplianceCertified DNA/RNase Free
ColorYellow
Volume (Metric) Working0.2 mL
Volume (Metric)0.25 mL
Quantity1000 tubes
For Use With (Equipment)Thermal Cyclers
MaterialPolypropylene
Reaction SpeedStandard
Closure TypeFlat Caps
Format1,000 Tubes
Product TypeTube
Unit Size1,000 tubes