You need to enable JavaScript to run this app.
Avidin, Egg White
Invitrogen™
Avidin, Egg White
Avidin from egg white is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used
to
Read more
Have Questions?
Change view
Catalog Number
Quantity
A887
100 mg
A2667
5 mg
Catalog number
A887
Price (USD)
/ 100 mg
560.00
-
Add to cart
Quantity:
100 mg
Price (USD)
/ 100 mg
560.00
Add to cart
Avidin, Egg White
Catalog number
A887
Price (USD)
/ 100 mg
560.00
-
Add to cart
Avidin from egg white
is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Conjugate
Unconjugated
Quantity
100 mg
Shipping Condition
Room Temperature
Unit Size
100 mg
Contents & Storage
Store in freezer (-5 to -30°C).