Catalog Number | Format | Quantity |
---|---|---|
A5757204 | BPC | 10 L |
A5757201 | Bottle | 1 L |
A5757202 | BPC | 1 L |
A5757203 | BPC | 5 L |
Benefits
• Provides enhanced T-cell expansion and expression of early memory phenotype makers compared to other chemically defined media
• Demonstrates flexibility across culture vessels. In both G-Rex™ plates and rocking bioreactors, it supports scale-up while maintaining desired T-cell phenotype.
• Expands both healthy and disease state donors
• Supports efficient viral and non-viral editing with downstream expansion
• Mitigates risk of contaminants and variability associated with animal-sourced materials
• Supports scale-up via larger catalog pack sizes
Features
• Animal-origin-free (AOF) formulation
• Serum-free—designed without the need for serum or serum replacement
• No separate components—no need to mix basal medium with supplement to obtain complete medium
• Yields high density cultures
• Supports T-cell activation using CTS Dynabeads (both passive release CD3/CD2 (Cat. No.40203D) and detachable CD3/CD28 (Cat No. A56992))
• Available in 1L bottle, 1L bag, 5L bag, and 10L bag options
Increased efficiency and consistent performance
CTS OpTmizer One SFM is an AOF formulation that supports high density cultures in static and dynamic vessels. It displays consistent performance for T-cell expansion, with better early memory phenotype at early stages.
Simplify your transition into clinical research
The Gibco Cell Therapy Systems (CTS) portfolio offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:
• Reduced burden in qualifying reagents
• Appropriate certificates of analysis and origin available
• Convenient access to our Drug Master File (DMF)