Absolute Q™ dPCR SARS-CoV-2 Wastewater Surveillance Kit
Absolute Q™ dPCR SARS-CoV-2 Wastewater Surveillance Kit
Applied Biosystems™

Absolute Q™ dPCR SARS-CoV-2 Wastewater Surveillance Kit

The Applied Biosystems Absolute Q dPCR SARS-CoV-2 Wastewater Surveillance Kit enables fast, sensitive, absolute quantification of SARS-CoV-2 from isolated wastewaterRead more
Have Questions?
Catalog number A55241
Price (USD)/ 200 reactions
2,258.65
Online exclusive
2345.00 
Save 86.35 (4%)
-
Add to cart
Price (USD)/ 200 reactions
2,258.65
Online exclusive
2345.00 
Save 86.35 (4%)
Add to cart
Absolute Q™ dPCR SARS-CoV-2 Wastewater Surveillance Kit
Catalog numberA55241
Price (USD)/ 200 reactions
2,258.65
Online exclusive
2345.00 
Save 86.35 (4%)
-
Add to cart
The Applied Biosystems Absolute Q dPCR SARS-CoV-2 Wastewater Surveillance Kit enables fast, sensitive, absolute quantification of SARS-CoV-2 from isolated wastewater nucleic acid using the QuantStudio Absolute Q Digital PCR System.

Features of the Absolute Q dPCR SARS-CoV-2 wastewater surveillance kit include:
Ease of use—simple qPCR-like workflow with just a few pipetting steps and minimal hands-on time
Fast—absolute quantification of targets of interest in less than two hours, including reaction preparation and dPCR data collection
Consistency—powered by microfluidic array plate (MAP) technology and TaqMan chemistry for exceptional data consistency

The Absolute Q SARS-CoV-2 Wastewater Surveillance Kit contains both a triplex digital PCR (dPCR) assay and Absolute Q 1-Step RT-dPCR Master Mix. The high-resolution multiplex assay targets two SARS-CoV-2 N gene (nucleoplasmid protein) regions because the gene is the more stable and less susceptible to mutations. The assay was designed by our experts with experience in rapidly responding to the SARS-CoV-2 outbreak, who helped ensure SARS-CoV-2 specificity and followed CDC guidance.

In accordance with CDC guidelines for SARS-CoV-2 wastewater surveillance, an assay targeting pepper mild mottle virus (PMMoV) is also part of the single tube triplex assay to enable fecal normalization. The assay enables all three targets to be detected in a single well. With the QuantStudio Absolute Q dPCR System, the red channel is free to enable further multiplexing with your own target of interest by combining the assay kit with an appropriate dye-labeled assay.

The wastewater surveillance kit offers enough multiplex assay and master mix for 200 sample reactions and is for use with the QuantStudio Absolute Q MAP16 Plate.

Related products
QuantStudio Absolute Q dPCR System
QuantStudio Absolute Q MAP16 Plate kit
MagMAX Wastewater Ultra Nucleic Acid Isolation Kit
MagMAX Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment
AcroMetrix SARS-CoV-2 Control (RUO)
AcroMetrix Coronavirus 2019 (COVID-19) RNA Control (RUO)

For Research Use Only.
Specifications
FormatTube
PCR MethodDigital PCR
Reaction Speed<2 hr.
TechniqueDigital PCR
For Use With (Equipment)QuantStudio™ Absolute Q™ Digital PCR System
Label or DyeFAM, VIC, ABY
Quantity200 reactions
Shipping ConditionDry Ice
TargetSARS-CoV-2 Pepper Mild Mottle Virus (PMMoV)
For Use With (Application)Pathogen Detection
Hot StartHot Start Taq Polymerase
No. of Reactions200
Unit Size200 reactions
Contents & Storage
• Absolute Q 1-step RT-dPCR Master Mix (4X)
• Absolute Q Wastewater Surveillance Assay (20X)

Store at –15 to –25°C.