CTS™ HiFi Cas9 Protein
TEG-Seq CRISPR Off-Target Analysis Service is available to help you monitor and detect off-target effects in your experiments. Learn more
CTS™ HiFi Cas9 Protein
Gibco™

CTS™ HiFi Cas9 Protein

Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining highRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A542245 mg
A542232.5 mg
Catalog number A54224
Price (USD)/ 5.0 mg
-
Request A Quote
Quantity:
5 mg

Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining high on-target activity. This protein is designed and manufactured in compliance with the principles of GMP and to meet requirements of the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products. It is provided in a high-concentration and large-scale format for process development and clinical research applications.

Features of CTS HiFi Cas9 Protein include:
Exceptional off-target profile—over 70% reduction in off-target effects in primary immune cells, compared to CTS TrueCut Cas9 (wild type) at tested loci
Maintain on-target activity—achieve comparable on-target activity to CTS TrueCut Cas9 (wild type) in primary immune cells at tested loci
Stringent manufacturing process—manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products including USP , Ph.Eur. 5.2.12, and ISO 20399 following the principles of 21 CFR Part 820 in an FDA-registered manufacturing site
Extensive safety testing—includes sterility, endotoxin, mycoplasma, residual nucleic acid, and protein testing
Traceability documentation—COA, COO, Drug Master files, and Regulatory Support files* are available

CTS HiFi Cas9 Protein is a Streptococcus pyogenes high-fidelity Cas9 protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR-Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.

If you would like to evaluate our HiFi Cas9 protein or conduct small-scale research experiments, we recommend using our research-grade TrueCut HiFi Cas9 Protein.


*Country-specific Regulatory Support File process will be initiated upon request and will require additional documentation time.

For Research Use or Non-Commercial Manufacturing of Cell-Based Products for Clinical Research. Caution: Not intended for direct administration into humans or animals.
Specifications
Concentration10 mg/mL
Quantity5 mg
Shelf Life18 months from date of manufacture
Product TypeCas9 Protein
Shipping ConditionDry Ice
Expression SystemE. coli
FormLiquid
Research CategoryGenome Editing
Manufacturing QualityISO 13485
Unit Size5.0 mg
Contents & Storage
Store at –20°C.