MagMAX™ Dx Viral/Pathogen NA Isolation Kit
MagMAX™ Dx Viral/Pathogen NA Isolation Kit
Applied Biosystems™

MagMAX™ Dx Viral/Pathogen NA Isolation Kit

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is specifically designed to recover viral and bacterial nucleic acids fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A52736Up to 600 preps
A52735Up to 2,000 preps
Catalog number A52736
Price (USD)/ Up to 600 preps
1,104.00
Add to cart
No. of Reactions:
Up to 600 preps
Price (USD)/ Up to 600 preps
1,104.00
Add to cart
MagMAX™ Dx Viral/Pathogen NA Isolation Kit
Catalog numberA52736
Price (USD)/ Up to 600 preps
1,104.00
-
Add to cart
The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is specifically designed to recover viral and bacterial nucleic acids from human respiratory biological specimens for qPCR downstream applications. The kit uses MagMAX magnetic-bead technology to enable reproducible recovery of high-quality nucleic acids.

Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• For in vitro diagnostic use. This product is US IVD compliant
• For isolation and purification of viral and bacterial nucleic acids from a range of respiratory biological specimens such as nasopharyngeal swab, anterior nasal swab, sputum, and saliva
• Single, flexible automation protocol that accommodates sample volume inputs of 200–400 μL
• Fast, automated procedures that allow 96 samples to be processed on the KingFisher Apex Dx Purification Instrument with ∼30-minute instrument run time

Access the Instructions for Use
(You will be redirected to sign into the resource library.)
Access the MagMAX Dx protocol for KingFisher Apex Dx instrument: Dx_MVP_RSP_APX_V1.kfdx
(You will be redirected to sign into the resource library.)
More information about the KingFisher Apex Dx Purification Instrument

Please check with your country specific representative for more information on product registration.

For In Vitro Diagnostic Use
Specifications
Format96-well Plate
Isolation TechnologyMagnetic Bead
Sample TypeNasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
For Use With (Equipment)KingFisher™ Apex Dx
For Use With (Application)Real-Time PCR
No. of ReactionsUp to 600 preps
Nucleic Acid TypeViral and Bacterial
Product LineMagMAX™
Shipping ConditionRoom Temperature
Shelf Life24 Months
TargetRespiratory Pathogens
Unit SizeUp to 600 preps
Contents & Storage

• Binding Solution, 160 mL
• Wash Solution, 300 mL
• Binding Beads, 6 mL
• Proteinase K, 3 mL
• Elution Buffer, 36 mL

Shipped and stored at room temperature.