Specifications
DescriptionProtein A/G Magnetic Beads
Kit ContentsTrypsin Protease, 2 x 20 μg
Trypsin Storage Solution, 250 μL
Protein A/G Magnetic Beads, 1 mL
IP-MS Cell Lysis Buffer, 100 mL
IP-MS Wash Buffer A, 75 mL
IP-MS Wash Buffer B, 40 mL
IP Elution and MS Sample Prep Buffer, 10 mL
Alkylation Solution, 1 mL/vial
Low Protein-Binding Collection Tubes, 50 x 1.5 mL
Bond-Breaker™ TCEP Solution, Neutral pH, 0.5 mL
10% Pierce™ Trifluoroacetic Acid (TFA), Sequencing Grade, 0.5 mL
1M MgCl2, 0.75 mL
For Use With (Equipment)Mass spectrometer
Product LineSureQuant™
Starting MaterialCells, Tissue
Detection MethodMass Spectrometry
Workflow StepSample prep enrichment
Quantity20 Reactions
For Use With (Application)MS-compatible immunoprecipitation
Unit Size20 rxns