Platinum™ SuperFi™ U Multiplex Master Mix
This product will be discontinued on April 30th, 2025. If you have any questions, please contact Customer Support.
Platinum™ SuperFi™ U Multiplex Master Mix
Invitrogen™

Platinum™ SuperFi™ U Multiplex Master Mix

Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Samples
A514002424 preps
A514009696 preps
Catalog number A5140024
Price (USD)/ 24 reactions
89.25
-
Add to cart
No. of Samples:
24 preps
Price (USD)/ 24 reactions
89.25
Add to cart
Platinum™ SuperFi™ U Multiplex Master Mix
Catalog numberA5140024
Price (USD)/ 24 reactions
89.25
-
Add to cart
Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use in next-generation sequencing. The ready-to-use 4X master mix contains uracil-tolerant high-fidelity DNA polymerase with all necessary buffer components and dNTPs for efficient targeted amplification.

Features of Platinum SuperFi U Multiplex Master Mix include:
• Optimized for high multiplex PCR
• Enables fast, efficient and robust amplification
• Provides high yields

Specifications
ColorTransparent
Concentration4X
EnzymePlatinum™ SuperFi™ U DNA Polymerase
For Use With (Application)Sars-Covid-2 Sequencing
No. of Reactions24 Preps
No. of Samples24 preps
Product LineCollibri™
Product TypeMaster Mix
SpeciesAll
Unit Size24 reactions
Contents & Storage
Store at -25°C to -15°C