Catalog Number | Gel Percentage | Separation Range | Quantity |
---|---|---|---|
A42390 | 2% | 50 to 2000 bp | 2 x 10 Gels |
A42347 | 1% | 100 to 5000 bp | 10 Gels |
A44885 | 1% | 100 to 5000 bp | 2 x 10 Gels |
A44886 | 1% | 100 to 5000 bp | 5 x 10 Gels |
A42348 | 2% | 50 to 2000 bp | 10 Gels |
A44884 | 2% | 50 to 2000 bp | 5 x 10 Gels |
Features of E-Gel Double Comb Agarose Gels include:
Less hazardous option
Choose SYBR Safe DNA gel stain to minimize exposure to hazardous and mutagenic options such as ethidium bromide and UV light. Precast E-Gel Double Comb gels with SYBR Safe stain, in combination with blue-light trans-illumination, offer better sample integrity and a less hazardous work environment.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng of sample DNA per band and a separation time 2–3 fold faster than that of conventional pour-your-own gels. Each E-Gel Double Comb gel contains agarose, electrodes, SYBR Safe DNA stain, and a buffer-less ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.
Live monitoring of DNA migration
E-Gel Double Comb gels include SYBR Safe DNA Gel Stain, which has an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, the gels enable less hazardous real-time monitoring of DNA migration without risk to the eyes or risk of damaging the sample, as opposed to UV visualization.
Easy access to the gel
E-Gel Double Comb cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for southern blot analysis.
Running and imaging devices
E-Gel Double Comb gels require the E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 22-well E-Gel Double Comb gels with SYBR Safe stain are available in 1% and 2% gel percentages.