Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Thermo Scientific™

Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)

Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven HeavyPeptide AQUA Ultimate peptides provided at five dilutions distinguished by differential isotopic labeling to assess sensitivity and dynamic range of LC-MS/MS systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
A400100.5 pmol/μL
A517475 pmol/μL
Catalog number A40010
Price (USD)/ 25 µL
186.65
Online exclusive
208.00 
Save 21.35 (10%)
-
Add to cart
Concentration:
0.5 pmol/μL
Price (USD)/ 25 µL
186.65
Online exclusive
208.00 
Save 21.35 (10%)
Add to cart
Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Catalog numberA40010
Price (USD)/ 25 µL
186.65
Online exclusive
208.00 
Save 21.35 (10%)
-
Add to cart

The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven PRTC HeavyPeptide AQUA Ultimate peptides in five isotopologue versions for the generation of seven standard curves to assess sensitivity and dynamic range of LC-MS/MS systems.

The Pierce LC-MS/MS System Suitability Standard (7 x 5 mix) contains 7 of 15 peptides from the Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88320). Each of the seven peptides has five versions (amino acids labeled with 0, 1, 2, 3, or 4 heavy isotopes) at five distinct concentrations. The 7 x 5 mix can be used with Skyline software to assess dynamic range, linearity, and lower limit of quantitation (LLOQ).

The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) is:
High quality—contains five sets of seven HeavyPeptide AQUA Ultimate peptides, each distinguishable through unique isotopologue labeling
Optimized—enables scheduling of MS acquisition windows for improved quantification and increased multiplexing
Quantitative—assesses dynamic range of instrument and instrument sensitivity
Predictive—enables assessment of chromatography and MS instrument performance

Applications:
• Optimization of LC & MS parameters
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., parallel-reaction monitoring (PRM)
• Identification of total peptide elution window
• Assessment dynamic range of nano or capillary flow LC-MS/MS systems

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture) (0.5 pmol/μL)
Concentration0.5 pmol/μL
Quantity25 μL
TypeLC-MS/MS System Standard
For Use With (Equipment)Mass Spectrometer
Product LinePierce™
Shipping ConditionDry Ice
Unit Size25 µL