EasyPep™ MS Sample Prep Kits
EasyPep™ MS Sample Prep Kits
Thermo Scientific™
EasyPep™ MS Sample Prep Kits
Generate high quality proteomic samples for MS analysis in less than four hours using optimized EasyPep MS Sample Prep kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
A40006Mini MS Sample Prep Kit
A57864Micro MS Sample Prep Kit
A45733MS Sample Prep Kit
A45734Maxi Sample Prep Kit
A45735Lysis Buffer
A57865Peptide Clean-up Plate
Catalog number A40006
Price (USD)/ 20 reactions
602.65
Online exclusive
652.00 
Save 49.35 (8%)
-
Add to cart
Description:
Mini MS Sample Prep Kit
Price (USD)/ 20 reactions
602.65
Online exclusive
652.00 
Save 49.35 (8%)
Add to cart
EasyPep™ MS Sample Prep Kits
Catalog numberA40006
Price (USD)/ 20 reactions
602.65
Online exclusive
652.00 
Save 49.35 (8%)
-
Add to cart
EasyPep Mini MS Sample Prep Kits enable efficient and reproducible processing of cultured mammalian cells and tissues for proteomic MS analysis. The kit contains pre-formulated buffers, MS-grade enzyme mix, peptide clean-up columns, and an optimized protocol to generate MS-compatible peptide samples in less than three hours. Kits are optimized to process protein samples from 10–100 μg with a high yield of MS-ready peptides.
Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
Time-saving—sample processing time reduced from more than one day to less than three hours
Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling
The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMini MS Sample Prep Kit
For Use With (Equipment)Mass Spectrometer
Product TypeSample Preparation Kit
Product LineEasyPep™
Shipping ConditionWet Ice
Starting MaterialProtein Samples
Final Product TypePeptides
Detection MethodMass Spectrometry
Workflow StepCell Lysis
Quantity20 Reactions
Unit Size20 reactions