Aluora™ Spatial Amplification Reagent Components
Aluora™ Spatial Amplification Reagent Components
Invitrogen™

Aluora™ Spatial Amplification Reagent Components

Invitrogen Aluora spatial amplification reagent components provide the critical reagents for use with Aluora spatial amplification dyes and kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
A40002450Spatial Rainbow Kit with Prolong Glass800 Reactions
AS100HRPSpatial Amplification Core Reagent Kit200 Reactions
AS201HRPGoat anti-Mouse (GAM) PolyHRP Secondary Reagent200 Reactions
AS202HRPGoat anti-Rabbit (GAR) PolyHRP Secondary Reagent200 Reactions
AS203HRPStreptavidin (SA) HRP Secondary Reagent200 Reactions
Catalog number A40002450
Price (USD)/ 800 reactions
6,900.00
-
Add to cart
Product Type:
Spatial Rainbow Kit with Prolong Glass
Quantity:
800 Reactions
Price (USD)/ 800 reactions
6,900.00
Add to cart
Aluora™ Spatial Amplification Reagent Components
Catalog numberA40002450
Price (USD)/ 800 reactions
6,900.00
-
Add to cart
Invitrogen Aluora spatial amplification reagent components provide the critical reagents for use with Aluora spatial amplification dyes and kits.

Revolutionary signal amplification technology

The Aluora spatial amplification kits represent a breakthrough in signal amplification for spatial proteomics. They employ proprietary Aluora reagents that react with horseradish peroxidase (HRP) to deposit photostable Aluora dyes onto target proteins, significantly improving the signal-to-background ratio. This technology enables robust multiplexed protein detection even in challenging tissue samples. Aluora kits require 10–5000X less primary antibody than standard IHC to achieve equivalent signal intensity.

Aluora spatial amplification reagents are optimized for use with Prolong Glass Antifade mountants and 1 mg/mL Thermo Scientific DAPI nucleic acid stains for multiplex analysis.

Aluora reagents and dyes can be purchased individually or in pre-configured kits to detect mouse, rabbit, or biotin-labelled antibodies. The Rainbow kit is configured with all Aluora colors with 4X GAM and 4X GAR secondary detection core reagents for 100 slides in 8-plex.

Features of Aluora spatial amplification reagents include:

  • Signal amplification for enhanced detection sensitivity
  • Comprehensive spatial analysis and imaging capabilities for research
  • Fluorescent detection system for multiplexing and quantitative analysis
  • Flexible workflow options that accommodate diverse experimental needs
  • Polymerase HRP (PolyHRP) conjugated secondary antibodies that boost signal for low abundance targets
  • Highly cross-adsorbed secondary antibodies that help ensure target-specific detection and background reduction
  • Enhanced background reduction through proprietary blocking reagents for crisp, clear imaging

Optimized for automated slide staining

A major benefit of Aluora spatial amplification kits is their compatibility with automated slide stainers right out of the box. This enables complete labeling of a tissue slide with an 8-plex Aluora panel in just 12–16 hours.

Eliminating background for cleaner images

Aluora spatial amplification kits include blockers to help reduce endogenous peroxidase activity and autofluorescence. By minimizing background noise, less primary antibody is needed to generate a strong specific signal.

Iterative labeling enables unlimited multiplexing

The covalently bound Aluora dyes are resistant to antibody stripping, remaining brightly fluorescent through subsequent labeling cycles. This enables iterative staining of numerous targets using primary antibodies from the same host species—a major constraint in traditional immunohistochemistry. Any number of directly conjugated primary antibodies can also be introduced after Aluora labeling is complete.

Custom optimization for diverse applications

Although general protocols are provided, each sample requires optimization for optimal results. Tissue preparation and processing methods vary widely and can profoundly impact staining. Careful optimization of the complete workflow is critical to achieve the highest quality multiplexed data from precious samples. Aluora spatial amplification provides a versatile tool to address diverse spatial proteomics applications.

For Research Use Only.
Specifications
Product TypeSpatial Rainbow Kit with Prolong Glass
Target SpeciesMouse, Rabbit
Quantity800 Reactions
For Use With (Application)Spatial Proteomics, Immunohistochemistry (IHC), IHC Staining and Detection, Fluorescent Staining, Immunofluorescence (IF)
Product LineAluora™
Detection MethodFluorescence
Sample TypeFFPE Tissue
Kit ContentsAluora™ Dyes, GAM/GAR HRP, Prolong Glass, DAPI, Core reagents
Sufficient ForSpatial Proteomics
Excitation/Emission427/499, 493/518, 512/529, 553/567, 589/615, 652/670, 687/706, 757/783 nm
Unit Size800 reactions
Contents & Storage
  • 4 x 200 reactions Aluora Core Reagents Kits (2°C to 8°C Desiccate and protect from light)
  • 2 x 200 reactions Goat Anti-Mouse (GAM) PolyHRP Secondary Reagent (2°C to 8°C and Protect from light)
  • 2 x 200 reactions Goat Anti-Rabbit (GAR) PolyHRP Secondary Reagent (2°C to 8°C and Protect from light)
  • 1 x 100 reactions Aluora 430 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 488 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 514 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 555 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 594 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 647 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 700 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 1 x 100 reactions Aluora 750 Spatial Amplification Dye (-25°C to -5°C Desiccate and protect from light)
  • 2 x 2 mL Prolong Glass Antifade Mountant (Store at 2°C to 8°C. For long-term storage, store at -5° to -30°C and Avoid freeze/thaw cycles)
  • 1 x 1 mL DAPI (1mg/mL) (2°C to 8°C and Protect from light)