SGS™ All Species ID Food DNA Analyser Kits
SGS™ All Species ID Food DNA Analyser Kits
Actual product may vary
Thermo Scientific™

SGS™ All Species ID Food DNA Analyser Kits

Identify meat, plant or fish species present in all food sample types with ready-to-use DNA test kits, software and Ion Torrent NGS technology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionBarcode
A38453SGS All Species ID MEAT DNA Analyser Kit II25-48
A38452SGS All Species ID MEAT DNA Analyser Kit I 1-24
A38454SGS All Species ID FISH DNA Analyser Kit I1-24
A38455SGS All Species ID FISH DNA Analyser Kit II25-48
A38456SGS All Species ID PLANT DNA Analyser Kit I1-24
A38457SGS All Species ID PLANT DNA Analyser Kit II25-48
A38458SGS All Species ID Software
Catalog number A38453
Price (USD)/ Each
-
Request A Quote
Description:
SGS All Species ID MEAT DNA Analyser Kit II
Barcode:
25-48

Identify the species present in even the most challenging sample types with the Thermo Scientific™ Next Generation Sequencing (NGS) Food Authenticity Workflow incorporating SGS™ All Species ID Food DNA Analyser Kits.

Protect your business from food fraud by verifying the meat, plant or fish species in your food samples for improved traceability and food safety.

As part of a complete, simple Next Generation Sequencing workflow, these kits provide a list of meat, plant or fish species present in all types of food and feed products including complex and highly processed sample types.

Identification of the species present in food and feed samples is a critical step for proof of authenticity, verification of origin, traceability of raw materials and quality control of handling and cleaning processes in food production lines.

The Thermo Scientific NGS Food Authenticity Workflow utilizes the Ion Torrent™ Next Generation Sequencing technology to enable an untargeted screening approach making it possible to identify the species contained in a food sample by comparison with a DNA database of meat, plant or fish species.

The ready-to-use SGS All Species ID Food DNA Analyser Kits allow the unique labelling and amplification of meat, fish or plant DNA by PCR, thereby preparing the sample for Next Generation Sequencing, and automated data analysis by the SGS All Species ID Software. Test results provide a list of the meat, plant or fish species present in the sample.

  • Simple: ready-to-use kits include all necessary reagents for DNA labeling and amplification, and provide results with any sample type, including complex and processed samples.
  • Rapid: allows preparation of NGS library from extracted DNA within 4 hours.
  • Flexible: utilizes a unique labeling system in which the DNA of each food sample is individually identified with an exclusive barcoded DNA sequence, enabling you to analyze multiple samples together for meat, fish or plant species.  
  • Supported: our experienced technical team is ready to assist you.

Each SGS All Species ID Food DNA Analyser Kit is available in two different formats with 24 different barcodes each, enabling you to label up to 48 samples at once for multiple simultaneous sample analysis.

For Laboratory Use
For Laboratory Use
Specifications
Barcode25-48
DescriptionSGS All Species ID MEAT DNA Analyser Kit II
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Contents & Storage
≤-18°C