You need to enable JavaScript to run this app.
AutoLys M Tube Racks
Applied Biosystems™
AutoLys M Tube Racks
These AutoLys M Tube Racks hold 24 AutoLys M Tubes per rack. The racks are locked and unlocked using
the
Read more
Have Questions?
Catalog number
A37955
Price (USD)
/ 5 racks
130.65
Online exclusive
137.00
Save 6.35 (5%)
-
Add to cart
Price (USD)
/ 5 racks
130.65
Online exclusive
137.00
Save 6.35 (5%)
Add to cart
AutoLys M Tube Racks
Catalog number
A37955
Price (USD)
/ 5 racks
130.65
Online exclusive
137.00
Save 6.35 (5%)
-
Add to cart
These AutoLys M Tube Racks hold 24 AutoLys M Tubes per rack. The racks are locked and unlocked using the AutoLys M TubeLifter.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
AutoLys TubeLifter System
Quantity
5 racks
Shipping Condition
Room Temperature
Unit Size
5 racks
Contents & Storage
5 racks (each rack holds up to 24 Tubes)